Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-chlorophenyl)benzimidazole, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
3574-96-7
C13H9ClN2
228.68
OILIYWFQRJOPAI-UHFFFAOYSA-N
chlorfenazole,2-2-chlorophenyl benzimidazole,2-2-chlorophenyl-1h-benzimidazole,chlorfenazole iso,1h-benzimidazole, 2-2-chlorophenyl,2-2-chlorophenyl-1h-benzoimidazole,2-2-chlorophenyl-1h-benzo d imidazole,unii-rt668w2svp,2-o-chlorophenyl benzimidazole,2-2-chlorophenyl-1h-1,3-benzodiazole
77123
2-(2-chlorophenyl)-1H-benzimidazole
C1=CC=C(C(=C1)C2=NC3=CC=CC=C3N2)Cl
3574-96-7
C13H9ClN2
228.68
OILIYWFQRJOPAI-UHFFFAOYSA-N
chlorfenazole,2-2-chlorophenyl benzimidazole,2-2-chlorophenyl-1h-benzimidazole,chlorfenazole iso,1h-benzimidazole, 2-2-chlorophenyl,2-2-chlorophenyl-1h-benzoimidazole,2-2-chlorophenyl-1h-benzo d imidazole,unii-rt668w2svp,2-o-chlorophenyl benzimidazole,2-2-chlorophenyl-1h-1,3-benzodiazole
77123
2-(2-chlorophenyl)-1H-benzimidazole
C1=CC=C(C(=C1)C2=NC3=CC=CC=C3N2)Cl
Melting Point 223.2°C to 234.7°C
Infrared Spectrum Authentic
Quantity 5g
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 228.68
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-(2-chlorophenyl)benzimidazole
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning