Learn More
2,2-Di(tert-butylperoxy)butane, 50% solution in aromatic free mineral spirit, ACROS Organics™
Description
Specifications
CAS Min % | 49.0,49.0 |
CAS Max % | 51.0,51.0 |
Additional Information | hydroperoxides (as TBHP): < 0.3% |
Density | 0.8050g/mL |
Formula Weight | 234.34 |
Physical Form | Solution |
Percent Purity | 49 to 51% |
Packaging | Plastic bottle |
Solubility | Solubility in water: immiscible. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons |
Specific Gravity | 0.805 |
Show More |
Safety and Handling
GHS H Statement:
Flammable liquid and vapour.
Heating may cause a fire.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
May be fatal if swallowed and enters airways.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 218-507-2
TSCA : TSCA