Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Di(tert-butylperoxy)butane, 50% solution in aromatic free mineral spirit, ACROS Organics™

Click to view available options
Quantity:
10mL
500mL
Packaging:
Plastic bottle
2167-23-9,31807-55-3,78-93-3
C12H26O4
234.336
MFCD00048243
HQOVXPHOJANJBR-UHFFFAOYSA-N
2,2-di tert-butylperoxy butane,2,2-bis tert-butylperoxy butane,chaloxyd p 1293al,trigonox d,chaloxyd p 1200al,butane, 2,2-bis tert-butyldioxy,unii-v15k90hftj,v15k90hftj,2,2-bis t-butylperoxy butane,di-tert-butyl sec-butylidene diperoxide
221260
2,2-bis(tert-butylperoxy)butane
CCC(C)(OOC(C)(C)C)OOC(C)(C)C
2167-23-9,31807-55-3,78-93-3
C12H26O4
234.336
MFCD00048243
HQOVXPHOJANJBR-UHFFFAOYSA-N
2,2-di tert-butylperoxy butane,2,2-bis tert-butylperoxy butane,chaloxyd p 1293al,trigonox d,chaloxyd p 1200al,butane, 2,2-bis tert-butyldioxy,unii-v15k90hftj,v15k90hftj,2,2-bis t-butylperoxy butane,di-tert-butyl sec-butylidene diperoxide
221260
2,2-bis(tert-butylperoxy)butane
CCC(C)(OOC(C)(C)C)OOC(C)(C)C
Radical initiators
CAS Min % 49.0,49.0
CAS Max % 51.0,51.0
Additional Information hydroperoxides (as TBHP): < 0.3%
Density 0.8050g/mL
Formula Weight 234.34
Physical Form Solution
Percent Purity 49 to 51%
Packaging Plastic bottle
Solubility Solubility in water: immiscible. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons
Specific Gravity 0.805
Viscosity 2.16 mPa.s (20°C)
Color Undesignated
Quantity 500mL
Flash Point 43°C
Chemical Name or Material 2, 2-Di(tert-butylperoxy)butane, 50% Solution in Aromatic Free Mineral Spirit
Show More Show Less

GHS H Statement:
Flammable liquid and vapour.
Heating may cause a fire.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
May be fatal if swallowed and enters airways.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 218-507-2

TSCA : TSCA