Alfa Aesar™ 2,2'-Diaminodiphenyl disulfide, 97%

Manufacturer: Alfa Aesar™ H5580703

Quantity 1g

 View more versions of this product

Catalog No. AAH5580703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1141-88-4
    Molecular Formula C12H12N2S2
    Formula Weight 248.36
    MDL Number MFCD00007703
    Beilstein 914032
    Synonym 2,2'-Dithiodianiline; Bis(2-aminophenyl) disulfide
    Chemical Name or Material 2,2'-Diaminodiphenyl disulfide

    Specifications

    Formula Weight 248.36
    TSCA TSCA
    MDL Number MFCD00007703
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    Molecular Formula C12H12N2S2
    CAS 1141-88-4
    Beilstein 914032
    Synonym 2,2'-Dithiodianiline; Bis(2-aminophenyl) disulfide
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H318-H315-H335
    Toxic if swallowed.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.
    RTECS Number BX9540000
    Melting Point 91° to 92°C
    Chemical Name or Material 2,2'-Diaminodiphenyl disulfide
    Assay 97%

    Description

    Applications
    2,2 ft.-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.

    Solubility
    Soluble in hot methanol- almost transparent.

    Notes
    Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    GHS H Statement
    H301-H318-H315-H335
    Toxic if swallowed.
    Causes serious eye damage.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit