Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dibromobiphenyl, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13029-09-9
C12H8Br2
312.01
MFCD00093707
DRKHIWKXLZCAKP-UHFFFAOYSA-N
2,2'-dibromobiphenyl,2,2'-dibromo-1,1'-biphenyl,o,o'-dibromobiphenyl,1-bromo-2-2-bromophenyl benzene,1,1'-biphenyl, 2,2'-dibromo,unii-gsl06p6p2l,biphenyl, 2,2'-dibromo,gsl06p6p2l,1,1'-biphenyl, 2,2'-dibromo-9ci,biphenyl,2'-dibromo
83060
1-bromo-2-(2-bromophenyl)benzene
C1=CC=C(C(=C1)C2=CC=CC=C2Br)Br
13029-09-9
C12H8Br2
312.01
MFCD00093707
DRKHIWKXLZCAKP-UHFFFAOYSA-N
2,2'-dibromobiphenyl,2,2'-dibromo-1,1'-biphenyl,o,o'-dibromobiphenyl,1-bromo-2-2-bromophenyl benzene,1,1'-biphenyl, 2,2'-dibromo,unii-gsl06p6p2l,biphenyl, 2,2'-dibromo,gsl06p6p2l,1,1'-biphenyl, 2,2'-dibromo-9ci,biphenyl,2'-dibromo
83060
1-bromo-2-(2-bromophenyl)benzene
C1=CC=C(C(=C1)C2=CC=CC=C2Br)Br
Quantity 1g
Packaging Glass bottle
Melting Point 79°C to 83°C
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 312.01
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2, 2'-Dibromobiphenyl
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning