Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2-Dibromopropane, 95%

Click to view available options
Quantity:
1g
5g
594-16-1
C3H6Br2
201.889
MFCD00000119
ARITXYXYCOZKMU-UHFFFAOYSA-N
propane, 2,2-dibromo,2,2-dibromopropane,dibromo propane,2-dibrompropan,acmc-1aolk,ksc272i9f,inchi=1/c3h6br2/c1-3 2,4 5/h1-2h
11658
2,2-dibromopropane
CC(C)(Br)Br
594-16-1
C3H6Br2
201.889
MFCD00000119
ARITXYXYCOZKMU-UHFFFAOYSA-N
propane, 2,2-dibromo,2,2-dibromopropane,dibromo propane,2-dibrompropan,acmc-1aolk,ksc272i9f,inchi=1/c3h6br2/c1-3 2,4 5/h1-2h
11658
2,2-dibromopropane
CC(C)(Br)Br
Refractive Index 1.498
Quantity 1g
Color Colorless to Yellow
Density 1.85g/mL
Boiling Point 113°C to 114°C
Linear Formula (CH3)2CBr2
Beilstein 1731162
Formula Weight 201.89
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2,2-Dibromopropane
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA