Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dibromopropane, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass bottle
594-16-1
C3H6Br2
201.89
MFCD00000119
ARITXYXYCOZKMU-UHFFFAOYSA-N
propane, 2,2-dibromo,2,2-dibromopropane,dibromo propane,2-dibrompropan,acmc-1aolk,ksc272i9f,inchi=1/c3h6br2/c1-3 2,4 5/h1-2h
11658
2,2-dibromopropane
CC(C)(Br)Br
594-16-1
C3H6Br2
201.89
MFCD00000119
ARITXYXYCOZKMU-UHFFFAOYSA-N
propane, 2,2-dibromo,2,2-dibromopropane,dibromo propane,2-dibrompropan,acmc-1aolk,ksc272i9f,inchi=1/c3h6br2/c1-3 2,4 5/h1-2h
11658
2,2-dibromopropane
CC(C)(Br)Br
Boiling Point 114°C (740.0mmHg)
Flash Point >110°C
Specific Gravity 1.78
Density 1.7800g/mL
Refractive Index 1.4950 to 1.499
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 50g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CBr2CH3
Beilstein 01, 111
Formula Weight 201.89
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 2-Dibromopropane, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-828-9

TSCA : TSCA