Alfa Aesar™ 2,2-Dichloro-1,3-benzodioxole, 97%

Manufacturer: Alfa Aesar™ H3205603

Quantity 1g

 View more versions of this product

Catalog No. AAH3205603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2032-75-9
    Molecular Formula C7H4Cl2O2
    Formula Weight 191.01
    MDL Number MFCD00196818
    UN Number UN3265
    Chemical Name or Material 2,2-Dichloro-1,3-benzodioxole

    Specifications

    Formula Weight 191.01
    Flash Point 85°C (185°F)
    Quantity 1g
    Molecular Formula C7H4Cl2O2
    MDL Number MFCD00196818
    CAS 2032-75-9
    UN Number UN3265
    Boiling Point 215°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H314-H318-H227-H317
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 2,2-Dichloro-1,3-benzodioxole
    Assay 97%

    GHS H Statement
    H301-H314-H318-H227-H317
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit