Alfa Aesar™ (2,2-Dichloro-1-methylcyclopropyl)benzene, 95%

Manufacturer: Alfa Aesar™ B2557106

Quantity 5g

 View more versions of this product

Catalog No. AAB2557106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3591-42-2
    Molecular Formula C10H10Cl2
    Formula Weight 201.09
    MDL Number MFCD00012260
    Synonym 1,1-Dichloro-2-methyl-2-phenylcyclopropane
    Chemical Name or Material (2,2-Dichloro-1-methylcyclopropyl)benzene

    Specifications

    Formula Weight 201.09
    TSCA TSCA
    MDL Number MFCD00012260
    CAS 3591-42-2
    Boiling Point 70° to 72°C (0.8mmHg)
    Synonym 1,1-Dichloro-2-methyl-2-phenylcyclopropane
    Quantity 5g
    Molecular Formula C10H10Cl2
    Chemical Name or Material (2,2-Dichloro-1-methylcyclopropyl)benzene
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit