Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2,2-Dichlorocyclopropyl)benzene, 97%, Acros Organics

Click to view available options
Quantity:
25g
2415-80-7
C9H8Cl2
187.063
MFCD00001272
WLWFQGXZIDYWQF-UHFFFAOYSA-N
2,2-dichlorocyclopropyl benzene,1,1-dichloro-2-phenylcyclopropane,2,2-dichlorocyclopropyl-benzene,benzene, 2,2-dichlorocyclopropyl,cyclopropane, 1,1-dichloro, 2-phenyl,pubchem18679,acmc-1clka,ksc491m2n,wlwfqgxzidywqf-uhfffaoysa,2,2-dichloro-cyclopropyl-benzene
95349
(2,2-dichlorocyclopropyl)benzene
C1C(C1(Cl)Cl)C2=CC=CC=C2
2415-80-7
C9H8Cl2
187.063
MFCD00001272
WLWFQGXZIDYWQF-UHFFFAOYSA-N
2,2-dichlorocyclopropyl benzene,1,1-dichloro-2-phenylcyclopropane,2,2-dichlorocyclopropyl-benzene,benzene, 2,2-dichlorocyclopropyl,cyclopropane, 1,1-dichloro, 2-phenyl,pubchem18679,acmc-1clka,ksc491m2n,wlwfqgxzidywqf-uhfffaoysa,2,2-dichloro-cyclopropyl-benzene
95349
(2,2-dichlorocyclopropyl)benzene
C1C(C1(Cl)Cl)C2=CC=CC=C2
Density 1.2g/mL
Refractive Index 1.5504 to 1.5524 (20°C, 589nm)
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5C3H3Cl2
Formula Weight 187.07
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material (2, 2-Dichlorocyclopropyl)benzene, 97%
Show More Show Less