Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2-Diethoxyacetophenone, 96%

Manufacturer:  Alfa Aesar™ B2446436

Quantity 500g

 View more versions of this product

Catalog No. AAB2446436


  Check Availability
Add to cart

Specifications

Description

Applications
It is an effective photoinitiator for UV-curable acrylate-based coatings, adhesives and inks. used in UV-curable system. Particularly useful for photo curing clear acrylate-based formulations where non-yellowing is important. It is also recommended for pigmented inks where shelf life and color stability are essential.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

6175-45-7
208.26
Phenylglyoxal diethyl acetal
C12H16O3
MFCD00009659
2,2-Diethoxyacetophenone

Specifications

500g
C12H16O3
MFCD00009659
>110°C (230°F)
2,2-Diethoxyacetophenone
6175-45-7
208.26
131° to 134°C (10mmHg)
Phenylglyoxal diethyl acetal
96%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit