Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2-Diethoxyacetophenone, 96%

Click to view available options
Quantity:
25g
6175-45-7
C12H16O3
208.257
MFCD00009659
PIZHFBODNLEQBL-UHFFFAOYSA-N
2,2-diethoxyacetophenone,ethanone, 2,2-diethoxy-1-phenyl,phenylglyoxal diethyl acetal,2,2-diethoxy-1-phenylethan-1-one,alpha,alpha-diethoxyacetophenone,unii-w4bcz1mak3,2,2-diethoxy acetophenone,ccris 5222,benzoylformaldehyde diethyl acetal,glyoxal, phenyl-, 2-diethyl acetal
22555
2,2-diethoxy-1-phenylethanone
CCOC(C(=O)C1=CC=CC=C1)OCC
6175-45-7
C12H16O3
208.257
MFCD00009659
PIZHFBODNLEQBL-UHFFFAOYSA-N
2,2-diethoxyacetophenone,ethanone, 2,2-diethoxy-1-phenyl,phenylglyoxal diethyl acetal,2,2-diethoxy-1-phenylethan-1-one,alpha,alpha-diethoxyacetophenone,unii-w4bcz1mak3,2,2-diethoxy acetophenone,ccris 5222,benzoylformaldehyde diethyl acetal,glyoxal, phenyl-, 2-diethyl acetal
22555
2,2-diethoxy-1-phenylethanone
CCOC(C(=O)C1=CC=CC=C1)OCC
Applications
It is an effective photoinitiator for UV-curable acrylate-based coatings, adhesives and inks. used in UV-curable system. Particularly useful for photo curing clear acrylate-based formulations where non-yellowing is important. It is also recommended for pigmented inks where shelf life and color stability are essential.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Flash Point >110°C (230°F)
Boiling Point 131°C to 134°C (10mmHg)
Quantity 25g
Formula Weight 208.26
Percent Purity 96%
Chemical Name or Material 2,2-Diethoxyacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA