Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2-Diethoxypropane, Technical Grade

Click to view available options
Quantity:
5g
25g
100g
126-84-1
C7H16O2
132.203
MFCD00009224
FGQLGYBGTRHODR-UHFFFAOYSA-N
acetone diethyl acetal,propane, 2,2-diethoxy,acetone diethyl ketal,acetone, diethyl acetal,usaf do-44,unii-18i1qhh2k2,acetonediethylketal,acetone diethylacetal,diethoxypropane,propane,2-diethoxy
31361
2,2-diethoxypropane
CCOC(C)(C)OCC
126-84-1
C7H16O2
132.203
MFCD00009224
FGQLGYBGTRHODR-UHFFFAOYSA-N
acetone diethyl acetal,propane, 2,2-diethoxy,acetone diethyl ketal,acetone, diethyl acetal,usaf do-44,unii-18i1qhh2k2,acetonediethylketal,acetone diethylacetal,diethoxypropane,propane,2-diethoxy
31361
2,2-diethoxypropane
CCOC(C)(C)OCC
Flash Point 7°C (45°F)
Boiling Point 44°C to 46°C (60mmHg)
Quantity 5g
UN Number UN3271
Formula Weight 132.2
Grade Technical
Chemical Name or Material 2,2-Diethoxypropane

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

TSCA : TSCA