Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diethoxypropane, Technical Grade, Thermo Scientific™

Catalog Number p-7027352
Click to view available options
Quantity:
5 g
25 g
100 g
126-84-1
C7H16O2
132.203
MFCD00009224
FGQLGYBGTRHODR-UHFFFAOYSA-N
acetone diethyl acetal, propane, 2,2-diethoxy, acetone diethyl ketal, acetone, diethyl acetal, usaf do-44, unii-18i1qhh2k2, acetonediethylketal, acetone diethylacetal, diethoxypropane, propane,2-diethoxy
31361
2,2-diethoxypropane
CCOC(C)(C)OCC
126-84-1
C7H16O2
132.203
MFCD00009224
FGQLGYBGTRHODR-UHFFFAOYSA-N
acetone diethyl acetal, propane, 2,2-diethoxy, acetone diethyl ketal, acetone, diethyl acetal, usaf do-44, unii-18i1qhh2k2, acetonediethylketal, acetone diethylacetal, diethoxypropane, propane,2-diethoxy
31361
2,2-diethoxypropane
CCOC(C)(C)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 44°C to 46°C (60 mmHg)
Flash Point 7°C (45°F)
Quantity 5 g
UN Number UN3271
Formula Weight 132.2
Grade Technical
Chemical Name or Material 2,2-Diethoxypropane
Hazard Category Danger
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
TSCA TSCA
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit