Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Difluoro-1,3-benzodioxole, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
25g
1583-59-1
C7H4F2O2
158.10
MFCD00236217
DGCOGZQDAXUUBY-UHFFFAOYSA-N
2,2-difluorobenzo d 1,3 dioxole, 2,2-difluorobenzodioxole, 2,2-difluoro-2h-1,3-benzodioxole, 1,2-difluoromethylenedioxy benzene, 2,2-difluoro-benzodioxole, 2,2-difluoro-benzo 1,3 dioxole, 1,3-benzodioxole, 2,2-difluoro, pubchem17570, acmc-1bus1, ksc491c9d
74103
2,2-difluoro-2H-1,3-benzodioxole
FC1(F)OC2=CC=CC=C2O1
1583-59-1
C7H4F2O2
158.10
MFCD00236217
DGCOGZQDAXUUBY-UHFFFAOYSA-N
2,2-difluorobenzo d 1,3 dioxole, 2,2-difluorobenzodioxole, 2,2-difluoro-2h-1,3-benzodioxole, 1,2-difluoromethylenedioxy benzene, 2,2-difluoro-benzodioxole, 2,2-difluoro-benzo 1,3 dioxole, 1,3-benzodioxole, 2,2-difluoro, pubchem17570, acmc-1bus1, ksc491c9d
74103
2,2-difluoro-2H-1,3-benzodioxole
FC1(F)OC2=CC=CC=C2O1
Infrared Spectrum Authentic
NMR Consistent with structure (1H−NMR)
Boiling Point 129.0°C to 130.0°C
Specific Gravity 1.308
Density 1.3080g/mL
Flash Point 32°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 158.11
Percent Purity 97%
Chemical Name or Material 2,2-Difluoro-1,3-benzodioxole
Show More Show Less

GHS H Statement Flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 216-431-4

RUO – Research Use Only