Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
128073-33-6
C5H6F2O2
136.1
HLFLYOQLHYYNLT-UHFFFAOYSA-N
2,2-difluoro-1-methylcyclopropanecarboxylic acid,-2,2-difluoro-1-methylcyclopropanecarboxylic acid,+/--2,2-difluoro-1-methylcyclopropanecarboxylic acid,2,2-difluoro-1-methylcyclopropane carboxylic acid,2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid,2,2-difluoro-1-methyl-cyclopropanecarboxylic acid,2,2-difluoro-1-methylcyclopropane-carboxylic acid,-2,2-difluoro-1-methylcyclopropane-carboxylicacid,2,2-difluoro-1-methyl cyclopropane carboxylic acid,+/--2,2-difluoro-1-methylcyclopropane-carboxylicacid
14695229
2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
CC1(CC1(F)F)C(=O)O
128073-33-6
C5H6F2O2
136.1
HLFLYOQLHYYNLT-UHFFFAOYSA-N
2,2-difluoro-1-methylcyclopropanecarboxylic acid,-2,2-difluoro-1-methylcyclopropanecarboxylic acid,+/--2,2-difluoro-1-methylcyclopropanecarboxylic acid,2,2-difluoro-1-methylcyclopropane carboxylic acid,2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid,2,2-difluoro-1-methyl-cyclopropanecarboxylic acid,2,2-difluoro-1-methylcyclopropane-carboxylic acid,-2,2-difluoro-1-methylcyclopropane-carboxylicacid,2,2-difluoro-1-methyl cyclopropane carboxylic acid,+/--2,2-difluoro-1-methylcyclopropane-carboxylicacid
14695229
2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
CC1(CC1(F)F)C(=O)O
Quantity 5g
Melting Point 62.0°C to 65.0°C
Assay Percent Range 97%
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material (±)-2, 2-Difluoro-1-methylcyclopropanecarboxylic acid

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning