Alfa Aesar™ 2,2-Difluoro-2-(fluorosulfonyl)acetic acid, 94%

Manufacturer: Alfa Aesar™ H2711906

Quantity 5g

 View more versions of this product

Catalog No. AAH2711906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1717-59-5
    Molecular Formula FSO2CF2CO2H
    Formula Weight 178.09
    MDL Number MFCD01320807
    UN Number UN3265
    Chemical Name or Material 2,2-Difluoro-2-(fluorosulfonyl)acetic acid

    Specifications

    Formula Weight 178.09
    Molecular Formula FSO2CF2CO2H
    MDL Number MFCD01320807
    UN Number UN3265
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Flash Point >110°C (230°F)
    Boiling Point 153°C
    Quantity 5g
    CAS 1717-59-5
    Health Hazard 1 Danger
    Chemical Name or Material 2,2-Difluoro-2-(fluorosulfonyl)acetic acid
    Assay 94%

    Description

    Applications
    2,2-Difluoro-2-(fluorosulfonyl)acetic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important reagent in the synthesis of difluorocarbenes and difluorocyclopropanes.

    Solubility
    It is soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Water and Moisture sensitive.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit