Alfa Aesar™ 2,2-Difluoro-N-methoxy-N-methylacetamide, 95%

Manufacturer: Alfa Aesar™ H25833MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH25833MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 142492-01-1
    Molecular Formula C4H7F2NO2
    Formula Weight 139.1
    MDL Number MFCD06248152
    Synonym N-Methyl-N-methoxydifluoroacetamide
    Chemical Name or Material 2,2-Difluoro-N-methoxy-N-methylacetamide

    Specifications

    Formula Weight 139.1
    Molecular Formula C4H7F2NO2
    MDL Number MFCD06248152
    CAS 142492-01-1
    Synonym N-Methyl-N-methoxydifluoroacetamide
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 40° to 41°C (0.7mmHg)
    Quantity 250mg
    Chemical Name or Material 2,2-Difluoro-N-methoxy-N-methylacetamide
    Assay 95%

    GHS H Statement
    H301-H315-H319
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit