Alfa Aesar™ 2,2-Difluorocyclopropanecarboxylic acid, 95%

Manufacturer: Alfa Aesar™ H25752MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH25752MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 107873-03-0
    Molecular Formula C4H4F2O2
    Formula Weight 122.07
    MDL Number MFCD04115828
    UN Number UN3261
    Chemical Name or Material 2,2-Difluorocyclopropanecarboxylic acid

    Specifications

    Formula Weight 122.07
    Molecular Formula C4H4F2O2
    MDL Number MFCD04115828
    CAS 107873-03-0
    UN Number UN3261
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Boiling Point 70°C (7mmHg)
    Melting Point 55° to 60°C
    Quantity 250mg
    Chemical Name or Material 2,2-Difluorocyclopropanecarboxylic acid
    Assay 95%

    Description

    Applications
    2,2-Difluorocyclopropanecarboxylic acid derivative (i.e. 2,2-difluorocyclopropanecarbonyl chloride) is used in Friedel-Crafts reactions with various arenes to prepare aryl 2,2-difluorocyclopropyl ketones.

    Solubility
    Soluble in water.

    Notes
    Incompatible with strong oxidizing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit