Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2'-Dihydroxy-4,4'-dimethoxybenzophenone, 98%

Click to view available options
Quantity:
25g
131-54-4
C15H14O5
274.272
MFCD00009606
SODJJEXAWOSSON-UHFFFAOYSA-N
2,2'-dihydroxy-4,4'-dimethoxybenzophenone,bis 2-hydroxy-4-methoxyphenyl methanone,benzophenone-6,cyasorb uv 12,methanone, bis 2-hydroxy-4-methoxyphenyl,uvinul d 49,caswell no. 353c,unii-7813j9cs1g,2,2-dihydroxy-4,4-dimethoxybenzophenone,4,4'-dimethoxy-2,2'-dihydroxybenzophenone
8570
bis(2-hydroxy-4-methoxyphenyl)methanone
COC1=CC(=C(C=C1)C(=O)C2=C(C=C(C=C2)OC)O)O
131-54-4
C15H14O5
274.272
MFCD00009606
SODJJEXAWOSSON-UHFFFAOYSA-N
2,2'-dihydroxy-4,4'-dimethoxybenzophenone,bis 2-hydroxy-4-methoxyphenyl methanone,benzophenone-6,cyasorb uv 12,methanone, bis 2-hydroxy-4-methoxyphenyl,uvinul d 49,caswell no. 353c,unii-7813j9cs1g,2,2-dihydroxy-4,4-dimethoxybenzophenone,4,4'-dimethoxy-2,2'-dihydroxybenzophenone
8570
bis(2-hydroxy-4-methoxyphenyl)methanone
COC1=CC(=C(C=C1)C(=O)C2=C(C=C(C=C2)OC)O)O
Melting Point 133°C to 136°C
Quantity 25g
Beilstein 1887087
Merck Index 14,1099
Formula Weight 274.27
Percent Purity 98%
Chemical Name or Material 2,2'-Dihydroxy-4,4'-dimethoxybenzophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DJ0900000

TSCA : TSCA