Alfa Aesar™ 2,2'-Dimethoxy-1,1'-binaphthyl, 97%

Manufacturer: Alfa Aesar™ H5108606

Quantity 5g

 View more versions of this product

Catalog No. AAH5108606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2960-93-2
    Molecular Formula C22H18O2
    Formula Weight 314.38
    MDL Number MFCD00091146
    UN Number UN3077
    Chemical Name or Material 2,2'-Dimethoxy-1,1'-binaphthyl

    Specifications

    Formula Weight 314.38
    MDL Number MFCD00091146
    Quantity 5g
    Odor Odorless
    Molecular Formula C22H18O2
    CAS 2960-93-2
    UN Number UN3077
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P273-P305+P351+P338-P501a
    Wear protective gloves.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material 2,2'-Dimethoxy-1,1'-binaphthyl
    Assay 97%

    Description

    Applications
    2,2 ft.-Dimethoxy-1,1 ft.-binaphthyl is used as organic chemical synthesis intermediate.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P273-P305+P351+P338-P501a
    Wear protective gloves.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit