Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2'-Dimethoxy-1,1'-binaphthyl, 97%

Manufacturer:  Alfa Aesar™ H5108603

Quantity 1g

 View more versions of this product

Catalog No. AAH5108603

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
2,2 ft.-Dimethoxy-1,1 ft.-binaphthyl is used as organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions.
Description & Specifications

Chemical Identifiers

2960-93-2
314.38
UN3077
C22H18O2
MFCD00091146
2,2'-Dimethoxy-1,1'-binaphthyl

Specifications

2960-93-2
314.38
UN3077
Odorless
97%
C22H18O2
MFCD00091146
1g
2,2'-Dimethoxy-1,1'-binaphthyl

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P273-P305+P351+P338-P501a
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit