Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dimethoxy-1,1'-binaphthyl, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2960-93-2
C22H18O2
314.38
MFCD00091146
BJAADAKPADTRCH-UHFFFAOYSA-N
2,2'-dimethoxy-1,1'-binaphthalene,2,2'-dimethoxy-1,1'-binaphthyl,r-+-2,2'-dimethoxy-1,1'-binaphthyl,r-2,2'-dimethoxy-1,1'-binaphthyl,r-2,2'-dimethoxy-1,1'-binaphthalene,s-2,2'-dimethoxy-1,1'-binaphthalene,s---2,2'-dimethoxy-1,1'-binaphthalene,s---2,2'-dimethoxy-1,1'-binaphthyl,r-+-2,2'-dimethoxy-1,1'-binaphthalene,s-2,2'-dimethoxy-1,1'-binaphthyl
235616
2-methoxy-1-(2-methoxynaphthalen-1-yl)naphthalene
COC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)OC
2960-93-2
C22H18O2
314.38
MFCD00091146
BJAADAKPADTRCH-UHFFFAOYSA-N
2,2'-dimethoxy-1,1'-binaphthalene,2,2'-dimethoxy-1,1'-binaphthyl,r-+-2,2'-dimethoxy-1,1'-binaphthyl,r-2,2'-dimethoxy-1,1'-binaphthyl,r-2,2'-dimethoxy-1,1'-binaphthalene,s-2,2'-dimethoxy-1,1'-binaphthalene,s---2,2'-dimethoxy-1,1'-binaphthalene,s---2,2'-dimethoxy-1,1'-binaphthyl,r-+-2,2'-dimethoxy-1,1'-binaphthalene,s-2,2'-dimethoxy-1,1'-binaphthyl
235616
2-methoxy-1-(2-methoxynaphthalen-1-yl)naphthalene
COC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)OC
Packaging Glass bottle
Quantity 5g
Color White to Beige
Infrared Spectrum Authentic
Melting Point 198.0°C to 202.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone (warm),chloroform,thf (warm),,dichloromethane,insoluble in ethyl alcohol,hexane,methanol,,toluene
Formula Weight 314.38
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2, 2'-Dimethoxy-1, 1'-binaphthyl, 98%
Show More Show Less