Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dimethoxy-1,1'-binaphthyl, 98%, Thermo Scientific™

Catalog Number p-114144
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2960-93-2
C22H18O2
314.38
MFCD00091146
BJAADAKPADTRCH-UHFFFAOYSA-N
2,2'-dimethoxy-1,1'-binaphthalene, 2,2'-dimethoxy-1,1'-binaphthyl, r-+-2,2'-dimethoxy-1,1'-binaphthyl, r-2,2'-dimethoxy-1,1'-binaphthyl, r-2,2'-dimethoxy-1,1'-binaphthalene, s-2,2'-dimethoxy-1,1'-binaphthalene, s---2,2'-dimethoxy-1,1'-binaphthalene, s---2,2'-dimethoxy-1,1'-binaphthyl, r-+-2,2'-dimethoxy-1,1'-binaphthalene, s-2,2'-dimethoxy-1,1'-binaphthyl
235616
2,2'-dimethoxy-1,1'-binaphthalene
COC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OC
2960-93-2
C22H18O2
314.38
MFCD00091146
BJAADAKPADTRCH-UHFFFAOYSA-N
2,2'-dimethoxy-1,1'-binaphthalene, 2,2'-dimethoxy-1,1'-binaphthyl, r-+-2,2'-dimethoxy-1,1'-binaphthyl, r-2,2'-dimethoxy-1,1'-binaphthyl, r-2,2'-dimethoxy-1,1'-binaphthalene, s-2,2'-dimethoxy-1,1'-binaphthalene, s---2,2'-dimethoxy-1,1'-binaphthalene, s---2,2'-dimethoxy-1,1'-binaphthyl, r-+-2,2'-dimethoxy-1,1'-binaphthalene, s-2,2'-dimethoxy-1,1'-binaphthyl
235616
2,2'-dimethoxy-1,1'-binaphthalene
COC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1OC

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 198.0°C to 202.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone (warm),chloroform,thf (warm),,dichloromethane,insoluble in ethyl alcohol,hexane,methanol,,toluene
Formula Weight 314.38
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2'-Dimethoxy-1, 1'-binaphthyl, 98%
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit