Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethoxy-2-phenylacetophenone, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
250g
Packaging:
Glass bottle
24650-42-8
C16H16O3
256.29
MFCD00008475
KWVGIHKZDCUPEU-UHFFFAOYSA-N
2,2-dimethoxy-2-phenylacetophenone,benzil dimethyl ketal,kayacure bdmk,irgacure 651,ethanone, 2,2-dimethoxy-1,2-diphenyl,lucirin bdk,photomer 51,benzil dimethylketal,esacure kb 1,irgacure 621
90571
2,2-dimethoxy-1,2-diphenylethanone
COC(C1=CC=CC=C1)(C(=O)C2=CC=CC=C2)OC
24650-42-8
C16H16O3
256.29
MFCD00008475
KWVGIHKZDCUPEU-UHFFFAOYSA-N
2,2-dimethoxy-2-phenylacetophenone,benzil dimethyl ketal,kayacure bdmk,irgacure 651,ethanone, 2,2-dimethoxy-1,2-diphenyl,lucirin bdk,photomer 51,benzil dimethylketal,esacure kb 1,irgacure 621
90571
2,2-dimethoxy-1,2-diphenylethanone
COC(C1=CC=CC=C1)(C(=O)C2=CC=CC=C2)OC
Heavy Metals 50ppm max
Packaging Glass bottle
Loss on Drying 0.2% max. (75°C, 2 hrs)
Infrared Spectrum Authentic
Melting Point 64°C to 68°C
Quantity 5g
Color White to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H5COC(OCH3)2C6H5
Merck Index 14, 3392
Formula Weight 256.29
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 2, 2-Dimethoxy-2-phenylacetophenone, 99%
Show More Show Less

LIGHT SENSITIVE

EINECSNumber : 246-386-6

RTECSNumber : KM5775658

TSCA : TSCA