Alfa Aesar™ 2,2'-Dimethoxybiphenyl, 97%

Manufacturer: Alfa Aesar™ B2341114

Quantity 25g

 View more versions of this product

Catalog No. AAB2341114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4877-93-4
    Molecular Formula C14H14O2
    Formula Weight 214.26
    MDL Number MFCD00067073
    Chemical Name or Material 2,2'-Dimethoxybiphenyl

    Specifications

    Formula Weight 214.26
    Melting Point 152° to 156°C
    MDL Number MFCD00067073
    CAS 4877-93-4
    Quantity 25g
    Molecular Formula C14H14O2
    Chemical Name or Material 2,2'-Dimethoxybiphenyl
    Assay 97%

    Description

    Applications
    2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.

    Solubility
    Soluble in chloroform, methanol.

    Notes
    Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit