Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,2'-Dimethoxybiphenyl, 97%
Click to view available options
Quantity:
5g
25g
100g
Description
Applications
2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.
Solubility
Soluble in chloroform, methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
2,2-Dimethoxy-1,1-biphenyl is used in the synthesis of biphenyl scaffolds in the preparation of biphenyl-tetrathiafulvalene derivatives.
Solubility
Soluble in chloroform, methanol.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications
Quantity | 100g |
Melting Point | 152°C to 156°C |
Solubility Information | Soluble in chloroform,methanol. |
Formula Weight | 214.26 |
Percent Purity | 97% |
Chemical Name or Material | 2,2'-Dimethoxybiphenyl |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures