Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethoxypropane, +98%, ACROS Organics™

Click to view available options
Quantity:
500mL
1L
2.5L
10L
Packaging:
Glass bottle
Metal Drum
77-76-9
C5H12O2
104.15
HEWZVZIVELJPQZ-UHFFFAOYSA-N
acetone dimethyl acetal,propane, 2,2-dimethoxy,acetone dimethyl ketal,acetone, dimethyl acetal,acetone-dimethyl acetal,2,2-dimethoxy propane,acetone dimethylacetal,2,2-dimethyoxypropane,2,2-dimethoxy-propane,dimethoxypropan
6495
2,2-dimethoxypropane
CC(C)(OC)OC
77-76-9
C5H12O2
104.15
HEWZVZIVELJPQZ-UHFFFAOYSA-N
acetone dimethyl acetal,propane, 2,2-dimethoxy,acetone dimethyl ketal,acetone, dimethyl acetal,acetone-dimethyl acetal,2,2-dimethoxy propane,acetone dimethylacetal,2,2-dimethyoxypropane,2,2-dimethoxy-propane,dimethoxypropan
6495
2,2-dimethoxypropane
CC(C)(OC)OC
Alkylation reagent
Viscosity 0.452 mPa.s (25°C)
Density 0.8400g/mL
Specific Gravity 0.84
Water 0.5% max. (K.F.)
Refractive Index 1.3760 to 1.3780
Infrared Spectrum Authentic
Packaging Metal Drum
Color Colorless
Quantity 10L
Melting Point -47.0°C
Boiling Point 83.0°C
Flash Point −5°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (CH3)2C(OCH3)2
Beilstein 01,648
Fieser 01,268; 05,226; 13,199; 15,138
Solubility Information Solubility in water: 18 g/100g (25°C). Other solubilities: soluble in acetone, benzene, ccl4, ether,, n-heptane and methanol
Formula Weight 104.15
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material 2,2-Dimethoxypropane
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger