Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
2033-24-1
C6H8O4
144.13
MFCD00006638
GXHFUVWIGNLZSC-UHFFFAOYSA-N
meldrum's acid,isopropylidene malonate,meldrums acid,1,3-dioxane-4,6-dione, 2,2-dimethyl,sub-isopropyl malonate,2,2-dimethyl-1,3-dioxan-4,6-dione,cycl-isopropylidene malonate,cyclic isopropylidene malonate,malonic acid cyclic isopropylidene ester,meldrum acid
16249
2,2-dimethyl-1,3-dioxane-4,6-dione
CC1(OC(=O)CC(=O)O1)C
2033-24-1
C6H8O4
144.13
MFCD00006638
GXHFUVWIGNLZSC-UHFFFAOYSA-N
meldrum's acid,isopropylidene malonate,meldrums acid,1,3-dioxane-4,6-dione, 2,2-dimethyl,sub-isopropyl malonate,2,2-dimethyl-1,3-dioxan-4,6-dione,cycl-isopropylidene malonate,cyclic isopropylidene malonate,malonic acid cyclic isopropylidene ester,meldrum acid
16249
2,2-dimethyl-1,3-dioxane-4,6-dione
CC1(OC(=O)CC(=O)O1)C
Flash Point >365°C
Water 0.1% max. (K.F.)
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige-Brown to Brown-White
Melting Point 92°C to 96°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Fieser 08,313; 10,252; 11,311; 13,172
Merck Index 15, 5885
Solubility Information Solubility in water: 2.5g/100 mL (20°C). Other solubilities: slightly soluble in ethanol and chloroform
Formula Weight 144.13
Physical Form Crystals or Powder
Percent Purity 98%
Chemical Name or Material 2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
Show More Show Less

GHS H Statement
Causes skin irritation.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 217-992-8

RTECSNumber : JH0800000

TSCA : TSCA