Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethyl-1,3-propanediol, 99%, Thermo Scientific Chemicals

Catalog Number ac408550250
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
126-30-7
C5H12O2
104.15
MFCD00004685
SLCVBVWXLSEKPL-UHFFFAOYSA-N
neopentyl glycol, 2,2-dimethyl-1,3-propanediol, dimethylolpropane, neopentylene glycol, neopentanediol, neopentylglycol, 1,3-propanediol, 2,2-dimethyl, neol, hydroxypivalyl alcohol, dimethyltrimethylene glycol
31344
2,2-dimethylpropane-1,3-diol
CC(C)(CO)CO
126-30-7
C5H12O2
104.15
MFCD00004685
SLCVBVWXLSEKPL-UHFFFAOYSA-N
neopentyl glycol, 2,2-dimethyl-1,3-propanediol, dimethylolpropane, neopentylene glycol, neopentanediol, neopentylglycol, 1,3-propanediol, 2,2-dimethyl, neol, hydroxypivalyl alcohol, dimethyltrimethylene glycol
31344
2,2-dimethylpropane-1,3-diol
CC(C)(CO)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 0.1mg KOH/g max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point 122.0°C to 128.0°C
Color White
Boiling Point 208.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2C(CH3)2CH2OH
Quantity 25 g
Beilstein 01, IV, 2551
Fieser 05,259
Merck Index 15, 6543
Solubility Information Solubility in water: 830g/L (20°C). Other solubilities: freely soluble in alcohol and ether
Formula Weight 104.15
Percent Purity 99%
Physical Form Crystalline Chunks
Chemical Name or Material 2, 2-Dimethyl-1, 3-propanediol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
EINECSNumber 204-781-0
RTECSNumber TY5775000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit