Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethyl-3-pentanol 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
3970-62-5
C7H16O
116.204
MFCD00004566
HMSVXZJWPVIVIV-UHFFFAOYSA-N
2,2-dimethyl-3-pentanol,3-pentanol, 2,2-dimethyl,3-pentanol,2-dimethyl,3-pentanol,2,2-dimethyl,dsstox_cid_12813,dsstox_rid_79039,dsstox_gsid_32813,hmsvxzjwpviviv-uhfffaoysa,3-pentanol, 2,2-dimethyl-8ci 9ci
19871
2,2-dimethylpentan-3-ol
CCC(C(C)(C)C)O
3970-62-5
C7H16O
116.204
MFCD00004566
HMSVXZJWPVIVIV-UHFFFAOYSA-N
2,2-dimethyl-3-pentanol,3-pentanol, 2,2-dimethyl,3-pentanol,2-dimethyl,3-pentanol,2,2-dimethyl,dsstox_cid_12813,dsstox_rid_79039,dsstox_gsid_32813,hmsvxzjwpviviv-uhfffaoysa,3-pentanol, 2,2-dimethyl-8ci 9ci
19871
2,2-dimethylpentan-3-ol
CCC(C(C)(C)C)O
Color Colorless
Density 0.82g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4215 to 1.4235 (20°C, 589nm)
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C2H5CH(OH)C(CH3)3
Beilstein 01, I, 207
Formula Weight 116.2
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2, 2-Dimethyl-3-pentanol, 97%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning