Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2-Dimethyl-N-(2-pyridyl)propionamide, 97%

Click to view available options
Quantity:
5g
25g
86847-59-8
C10H14N2O
178.235
MFCD00460277
CGSPVYCZBDFPHJ-UHFFFAOYSA-N
2,2-dimethyl-n-pyridin-2-yl-propionamide,2-pivaloylamino pyridine,2,2-dimethyl-n-pyridin-2-yl propanamide,n-pyridin-2-yl pivalamide,n-boc-2-aminopyridine,2,2-dimethyl-n-2-pyridyl propanamide,2,2-dimethyl-n-2-pyridinyl propanamide,propanamide, 2,2-dimethyl-n-2-pyridinyl,pivaloylamino pyridine,n-o-pyridylpivalamide
1051519
2,2-dimethyl-N-pyridin-2-ylpropanamide
CC(C)(C)C(=O)NC1=CC=CC=N1
86847-59-8
C10H14N2O
178.235
MFCD00460277
CGSPVYCZBDFPHJ-UHFFFAOYSA-N
2,2-dimethyl-n-pyridin-2-yl-propionamide,2-pivaloylamino pyridine,2,2-dimethyl-n-pyridin-2-yl propanamide,n-pyridin-2-yl pivalamide,n-boc-2-aminopyridine,2,2-dimethyl-n-2-pyridyl propanamide,2,2-dimethyl-n-2-pyridinyl propanamide,propanamide, 2,2-dimethyl-n-2-pyridinyl,pivaloylamino pyridine,n-o-pyridylpivalamide
1051519
2,2-dimethyl-N-pyridin-2-ylpropanamide
CC(C)(C)C(=O)NC1=CC=CC=N1
Quantity 5g
Melting Point 71°C to 75°C
Beilstein 4178333
Formula Weight 178.23
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2,2-Dimethyl-N-(2-pyridyl)propionamide