Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylbutane 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
75-83-2
C6H14
86.178
MFCD00009321
HNRMPXKDFBEGFZ-UHFFFAOYSA-N
neohexane,butane, 2,2-dimethyl,hsdb 75,unii-07l56l3mp2,ccris 6019,2,2-dimethylbutyl,3,3-dimethylbutyl,butane,2-dimethyl,2,2-dimethyl-butane,3,3-dimethylbutan-2-yl
6403
2,2-dimethylbutane
CCC(C)(C)C
75-83-2
C6H14
86.178
MFCD00009321
HNRMPXKDFBEGFZ-UHFFFAOYSA-N
neohexane,butane, 2,2-dimethyl,hsdb 75,unii-07l56l3mp2,ccris 6019,2,2-dimethylbutyl,3,3-dimethylbutyl,butane,2-dimethyl,2,2-dimethyl-butane,3,3-dimethylbutan-2-yl
6403
2,2-dimethylbutane
CCC(C)(C)C
Refractive Index 1.3675 to 1.3695
Infrared Spectrum Authentic
Packaging Glass bottle
Density 0.6490g/mL
Color Colorless to Yellow
Melting Point -115.0°C
Quantity 500mL
Flash Point −48°C
Boiling Point 50.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C2H5C(CH3)3
Beilstein 01, 150
Formula Weight 86.18
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 2-Dimethylbutane, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-906-8

TSCA : TSCA