Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Dimethylbutyric acid, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
595-37-9
C6H12O2
116.16
MFCD00004200
VUAXHMVRKOTJKP-UHFFFAOYSA-N
2,2-dimethylbutyric acid,butanoic acid, 2,2-dimethyl,2,2-dimethyl-butanoic acid,2,2-dimethyl butyric acid,2,2-dimethyl butanoic acid,unii-ay606cn05o,butyric acid, 2,2-dimethyl,dimethyl ethyl acetic acid,2,2-dimethyl-butyric acid,.alpha.,.alpha.-dimethylbutyric acid
11684
CHEBI:38649
2,2-dimethylbutanoic acid
CCC(C)(C)C(=O)O
595-37-9
C6H12O2
116.16
MFCD00004200
VUAXHMVRKOTJKP-UHFFFAOYSA-N
2,2-dimethylbutyric acid,butanoic acid, 2,2-dimethyl,2,2-dimethyl-butanoic acid,2,2-dimethyl butyric acid,2,2-dimethyl butanoic acid,unii-ay606cn05o,butyric acid, 2,2-dimethyl,dimethyl ethyl acetic acid,2,2-dimethyl-butyric acid,.alpha.,.alpha.-dimethylbutyric acid
11684
CHEBI:38649
2,2-dimethylbutanoic acid
CCC(C)(C)C(=O)O
Specific Gravity 0.928
Density 0.9280g/mL
Color Colorless to Yellow
Quantity 25g
Flash Point 79°C
Packaging Glass bottle
Boiling Point 186.0°C to 187.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 116.16
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2, 2-Dimethylbutyric acid
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 209-865-