Alfa Aesar™ 2,2-Dimethylglutaric anhydride, 98%

Manufacturer: Alfa Aesar™ B2530306

Quantity 5g

 View more versions of this product

Catalog No. AAB2530306

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2938-48-9
    Molecular Formula C7H10O3
    Formula Weight 142.15
    MDL Number MFCD00006682
    Beilstein 117056
    Synonym 3,3-Dimethyldihydro-2H-pyran-2,6(3H)-dione
    Chemical Name or Material 2,2-Dimethylglutaric anhydride

    Specifications

    Formula Weight 142.15
    Molecular Formula C7H10O3
    Melting Point 35° to 38°C
    Boiling Point 270°C
    MDL Number MFCD00006682
    CAS 2938-48-9
    Beilstein 117056
    Quantity 5g
    Flash Point >110°C (230°F)
    Synonym 3,3-Dimethyldihydro-2H-pyran-2,6(3H)-dione
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2,2-Dimethylglutaric anhydride
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit