Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2-Dimethylsuccinic acid, 99%

Click to view available options
Quantity:
1g
5g
25g
597-43-3
C6H10O4
146.142
MFCD00004196
GOHPTLYPQCTZSE-UHFFFAOYSA-N
2,2-dimethylsuccinic acid,2,2-dimethyl-succinic acid,unii-mx6tvr2107,2,2-dimethyl-butanedioic acid,succinic acid, 2,2-dimethyl,butanedioic acid, 2,2-dimethyl,2,2-dimethylbutanedioate,2,2-dimethyl-butanedioicacid,2,2,-dimethylsuccinate,2,2-dimethyl succinate
11701
CHEBI:86537
2,2-dimethylbutanedioic acid
CC(C)(CC(=O)O)C(=O)O
597-43-3
C6H10O4
146.142
MFCD00004196
GOHPTLYPQCTZSE-UHFFFAOYSA-N
2,2-dimethylsuccinic acid,2,2-dimethyl-succinic acid,unii-mx6tvr2107,2,2-dimethyl-butanedioic acid,succinic acid, 2,2-dimethyl,butanedioic acid, 2,2-dimethyl,2,2-dimethylbutanedioate,2,2-dimethyl-butanedioicacid,2,2,-dimethylsuccinate,2,2-dimethyl succinate
11701
CHEBI:86537
2,2-dimethylbutanedioic acid
CC(C)(CC(=O)O)C(=O)O
Quantity 1g
Melting Point 139°C to 142°C
Beilstein 1764476
Formula Weight 146.14
Percent Purity 99%
Chemical Name or Material 2,2-Dimethylsuccinic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning