Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diphenylethanol, 97%, ACROS Organics™

Click to view available options
Quantity:
2g
10g
Packaging:
Glass bottle
1883-32-5
C14H14O
198.26
MFCD00004729
NYLOEXLAXYHOHH-UHFFFAOYSA-N
beta-phenylphenethyl alcohol,2,2-diphenyl ethanol,benzeneethanol, .beta.-phenyl,acmc-20aojg,2,2-diphenyl-ethanol,2,2-diphenyl-1-ethanol,2,2-diphenylethan-1-ol,benzeneethanol, b-phenyl,2,2-diphenylethanol,ksc539o6l
74662
2,2-diphenylethanol
C1=CC=C(C=C1)C(CO)C2=CC=CC=C2
1883-32-5
C14H14O
198.26
MFCD00004729
NYLOEXLAXYHOHH-UHFFFAOYSA-N
beta-phenylphenethyl alcohol,2,2-diphenyl ethanol,benzeneethanol, .beta.-phenyl,acmc-20aojg,2,2-diphenyl-ethanol,2,2-diphenyl-1-ethanol,2,2-diphenylethan-1-ol,benzeneethanol, b-phenyl,2,2-diphenylethanol,ksc539o6l
74662
2,2-diphenylethanol
C1=CC=C(C=C1)C(CO)C2=CC=CC=C2
Flash Point >110°C
Boiling Point 160°C to 162°C (5.0mmHg)
Color White
Quantity 10g
Melting Point 53°C to 56°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (C6H5)2CHCH2OH
Beilstein 06, II, 640
Formula Weight 198.26
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2, 2-Diphenylethanol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-543-6