Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diphenylethylamine 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3963-62-0
C14H15N
197.28
MFCD00008143
RXMTUVIKZRXSSM-UHFFFAOYSA-N
2,2-diphenylethylamine,2,2-diphenyl-ethylamine,1-amino-2,2-diphenylethane,2,2-diphenylethan-1-amine,chembl10780,pubchem7986,acmc-1cswz,2,2-diphenylethyl-amine,2,2-di phenyl ethanamine
77575
2,2-diphenylethanamine
C1=CC=C(C=C1)C(CN)C2=CC=CC=C2
3963-62-0
C14H15N
197.28
MFCD00008143
RXMTUVIKZRXSSM-UHFFFAOYSA-N
2,2-diphenylethylamine,2,2-diphenyl-ethylamine,1-amino-2,2-diphenylethane,2,2-diphenylethan-1-amine,chembl10780,pubchem7986,acmc-1cswz,2,2-diphenylethyl-amine,2,2-di phenyl ethanamine
77575
2,2-diphenylethanamine
C1=CC=C(C=C1)C(CN)C2=CC=CC=C2
Flash Point 110°C
Boiling Point 139°C to 140°C (1.2mmHg)
Melting Point 43°C to 49°C
Packaging Glass bottle
Color White to Yellow
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula (C6H5)2CHCH2NH2
Beilstein 12, 1327
Formula Weight 197.28
Physical Form Fused Solid
Percent Purity ≥95%
Chemical Name or Material 2, 2-Diphenylethylamine, 96%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 223-565-7

TSCA : TSCA