Alfa Aesar™ 2,2-Diphenylglycine, 98%

Manufacturer: Alfa Aesar™ B2265006

Quantity 5g

 View more versions of this product

Catalog No. AAB2265006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3060-50-2
    Molecular Formula C14H13NO2
    Formula Weight 227.26
    MDL Number MFCD00008048
    Synonym Aminodiphenylacetic acid
    Chemical Name or Material 2,2-Diphenylglycine

    Specifications

    Formula Weight 227.26
    Molecular Formula C14H13NO2
    MDL Number MFCD00008048
    CAS 3060-50-2
    Synonym Aminodiphenylacetic acid
    Melting Point ≈246°C (dec.)
    Quantity 5g
    Chemical Name or Material 2,2-Diphenylglycine
    Assay 98%

    Description

    Applications
    2,2-Diphenylglycine is used as a synthesis reagent for amination reactions. It acts as an intermediate in pharmaceutical and in organic synthesis.

    Solubility
    It is sparingly soluble in water.

    Notes
    Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit