Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2-Diphenylglycine, 98%

Click to view available options
Quantity:
5g
25g
100g
3060-50-2
C14H13NO2
227.263
MFCD00008048
YBONNYNNFBAKLI-UHFFFAOYSA-N
2,2-diphenylglycine,alpha,alpha-diphenylglycine,alpha-aminodiphenylacetic acid,glycine, 2,2-diphenyl,alpha-amino-alpha-phenylbenzeneacetic acid,.alpha.-aminodiphenylacetic acid,.alpha.,.alpha.-diphenylglycine,benzeneacetic acid, .alpha.-amino-.alpha.-phenyl,aminodiphenylacetic acid,phenytoin impurity c
18289
2-amino-2,2-diphenylacetic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(=O)O)N
3060-50-2
C14H13NO2
227.263
MFCD00008048
YBONNYNNFBAKLI-UHFFFAOYSA-N
2,2-diphenylglycine,alpha,alpha-diphenylglycine,alpha-aminodiphenylacetic acid,glycine, 2,2-diphenyl,alpha-amino-alpha-phenylbenzeneacetic acid,.alpha.-aminodiphenylacetic acid,.alpha.,.alpha.-diphenylglycine,benzeneacetic acid, .alpha.-amino-.alpha.-phenyl,aminodiphenylacetic acid,phenytoin impurity c
18289
2-amino-2,2-diphenylacetic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(=O)O)N
Applications
2,2-Diphenylglycine is used as a synthesis reagent for amination reactions. It acts as an intermediate in pharmaceutical and in organic synthesis.

Solubility
It is sparingly soluble in water.

Notes
Incompatible with oxidizing agents.
Formula Weight 227.26
Percent Purity 98%
Melting Point ~246°C (decomposition)
Quantity 25g
Chemical Name or Material 2,2-Diphenylglycine