Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dipyridyl, 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
500g
Packaging:
Glass bottle
366-18-7
C10H8N2
156.19
ROFVEXUMMXZLPA-UHFFFAOYSA-N
2,2'-bipyridine,2,2'-dipyridyl,2,2'-bipyridyl,bipyridine,2,2'-dipyridine,2-2-pyridyl pyridine,2,2'-bipyridin,alpha,alpha'-dipyridyl,alpha,alpha'-bipyridyl,2,2-bipyridyl
1474
CHEBI:30351
2-pyridin-2-ylpyridine
C1=CC=NC(=C1)C2=CC=CC=N2
366-18-7
C10H8N2
156.19
ROFVEXUMMXZLPA-UHFFFAOYSA-N
2,2'-bipyridine,2,2'-dipyridyl,2,2'-bipyridyl,bipyridine,2,2'-dipyridine,2-2-pyridyl pyridine,2,2'-bipyridin,alpha,alpha'-dipyridyl,alpha,alpha'-bipyridyl,2,2-bipyridyl
1474
CHEBI:30351
2-pyridin-2-ylpyridine
C1=CC=NC(=C1)C2=CC=CC=N2
Indicator
Viscosity 1.51 mm2/s (90°C)
Boiling Point 272.0°C to 273.0°C
Color White
Suitability (for iron) passes
Sulfated Ash 0.1% max.
Melting Point 69.0°C to 72.0°C
Quantity 10g
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point 121°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Beilstein 23,199
Fieser 16,25
Merck Index 15,1241
Solubility Information Solubility in water: slightly soluble (5.5g/L 22°C). Other solubilities: very soluble in ethanol (500g/L 25°C), ether,, benzene, chloroform and petroleum ether
Formula Weight 156.19
Physical Form Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material 2, 2'-Dipyridyl
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger