Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2'-Dipyridyl disulfide, 98%

Manufacturer:  Alfa Aesar™ A1111822

Quantity 100g

 View more versions of this product

Catalog No. AAA1111822


  Check Availability
Add to cart

Specifications

Description

Applications
2,2 ft.-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.

Solubility
Soluble in water, benzene, tetrahydrofuran, dichloromethane, acetonitrile and dimethyformamide.

Notes
Incompatible with oxidizing agents and metals.
Description & Specifications

Chemical Identifiers

2127-03-9
220.32
154629
2,2'-Dipyridyl disulfide
C10H8N2S2
MFCD00006287
2,2'-Dithiodipyridine

Specifications

210°C (410°F)
100g
98%
220.32
154629
2,2'-Dipyridyl disulfide
56° to 59°C
2127-03-9
C10H8N2S2
MFCD00006287
2,2'-Dithiodipyridine
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit