Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dithiosalicylic Acid 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
119-80-2
C14H10O4S2
306.34
MFCD00002465
LBEMXJWGHIEXRA-UHFFFAOYSA-N
2,2'-dithiodibenzoic acid,2,2'-dithiosalicylic acid,benzoic acid, 2,2'-dithiobis,2-2-carboxyphenyl disulfanyl benzoic acid,bis 2-carboxyphenyl disulfide,bis o-carboxylphenyl disulfide,2,2-dithiosalicylic acid,2,2'-dithiobisbenzoic acid,unii-58mgb5279d,2,2'-dithiodi benzoic acid
8409
2-[(2-carboxyphenyl)disulfanyl]benzoic acid
C1=CC=C(C(=C1)C(=O)O)SSC2=CC=CC=C2C(=O)O
119-80-2
C14H10O4S2
306.34
MFCD00002465
LBEMXJWGHIEXRA-UHFFFAOYSA-N
2,2'-dithiodibenzoic acid,2,2'-dithiosalicylic acid,benzoic acid, 2,2'-dithiobis,2-2-carboxyphenyl disulfanyl benzoic acid,bis 2-carboxyphenyl disulfide,bis o-carboxylphenyl disulfide,2,2-dithiosalicylic acid,2,2'-dithiobisbenzoic acid,unii-58mgb5279d,2,2'-dithiodi benzoic acid
8409
2-[(2-carboxyphenyl)disulfanyl]benzoic acid
C1=CC=C(C(=C1)C(=O)O)SSC2=CC=CC=C2C(=O)O
Melting Point 287°C to 290°C
Color Brown
Packaging Plastic bottle
Infrared Spectrum Authentic
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula S2(C6H4CO2H)2
Beilstein 10, 129
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in dichloromethane,ether,benzene and,acetone
Formula Weight 306.34
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 2'-Dithiosalicylic acid, 98%
Show More Show Less

EINECSNumber : 204-352-8

RTECSNumber : DG9660000

TSCA : TSCA