Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Ethoxyethoxy)ethanol, 99-100%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
111-90-0
111-90-0
Packaging Amber Glass Bottle
Water 0.002
Acidity 0.0001
Quantity 1 L
CAS Max % 1
Percent Purity 99 to 100%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.