Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Ethylenebis(1,3-dithiane), 98+%, Thermo Scientific™

Click to view available options
Quantity:
1g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

CAS 14947-53-6
Melting Point 132°C to 135°C
Molecular Formula C10H18S4
MDL Number MFCD00040467
Beilstein 1237944
Quantity 1g
Synonym 2,2'-ethylenebis 1,3-dithiane, 2,2'-ethylene di-1,3-dithiane, 2-2-1,3-dithian-2-yl ethyl-1,3-dithiane, 2,2'-ethane-1,2-diylbis 1,3-dithiane, acmc-1c6g9, 1,2-di 1,3-dithian-2-yl ethane, 1,3-dithiane,2,2'-1,2-ethanediyl bis
InChI Key RVIPSMFHRZBJRI-UHFFFAOYSA-N
SMILES C(CC1SCCCS1)C1SCCCS1
IUPAC Name 2-[2-(1,3-dithian-2-yl)ethyl]-1,3-dithiane
Molecular Weight (g/mol) 266.49
PubChem CID 84720
Formula Weight 266.51
Percent Purity ≥98%
Chemical Name or Material 2,2'-Ethylenebis(1,3-dithiane)
Show More Show Less

EINECSNumber : 239-022-2

TSCA : No

Recommended Storage : Ambient temperatures