Alfa Aesar™ 2-(2-Ethylhexyloxy)-5-methoxy-1,4-benzenediacetonitrile, 98%

Manufacturer: Alfa Aesar™ H5104003

Quantity 1g

 View more versions of this product

Catalog No. AAH5104003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 213749-91-8
    Molecular Formula C19H26N2O2
    Formula Weight 314.42
    MDL Number MFCD03701544
    UN Number UN3439
    Chemical Name or Material 2-(2-Ethylhexyloxy)-5-methoxy-1,4-benzenediacetonitrile

    Specifications

    Formula Weight 314.42
    Molecular Formula C19H26N2O2
    MDL Number MFCD03701544
    CAS 213749-91-8
    UN Number UN3439
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H302-H332-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Quantity 1g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 2-(2-Ethylhexyloxy)-5-methoxy-1,4-benzenediacetonitrile
    Assay 98%

    GHS H Statement
    H311-H302-H332-H315-H319-H335
    Toxic in contact with skin.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit