Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(2-Fluorophenyl)pyrrolidine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in freezer(-20°C). Keep container tightly closed. Store in a cool, dry and well-ventilated place.
2-(2-Fluorophenyl)pyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in freezer(-20°C). Keep container tightly closed. Store in a cool, dry and well-ventilated place.
Specifications
Color | Colorless |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 165.21 |
Percent Purity | 95% |
Physical Form | Liquid |
Chemical Name or Material | 2-(2-Fluorophenyl)pyrrolidine |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
storageNote1 | -30° to -10°C |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction