Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(2-Hydroxyethyl)pyridine, 99%

Manufacturer:  Alfa Aesar™ B2357936

Quantity 500g

 View more versions of this product

Catalog No. AAB2357936


  Check Availability
Add to cart

Specifications

Description

Applications
2-(2-Hydroxyethyl)pyridine is a metabolite of the anti-vertigo drug Betahistine. It acts as a reagent for protection of the 5 ft.-phosphate of nucleotides as 2-pyridylethyl (Pet) esters. It is used for carboxyl protection of amino acids. It is also used to cleave such groups as Boc and Fmoc, and also to hydrogenolysis. It was used to prepare mononuclear nickel(II) acetate.

Solubility
Soluble in Chloroform (Slightly), Methanol (Slightly).

Notes
Hygroscopic. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.
Description & Specifications

Chemical Identifiers

103-74-2
123.16
111205
2-(2-Hydroxyethyl)pyridine
C7H9NO
MFCD00006364
2-Pyridineethanol; 2-(2-Pyridyl)ethanol

Specifications

117°C (243°F)
−8°C
103-74-2
123.16
111205
2-(2-Hydroxyethyl)pyridine
235° to 238°C
500g
C7H9NO
MFCD00006364
2-Pyridineethanol; 2-(2-Pyridyl)ethanol
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UT2970450

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit