Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Methoxyethoxy)ethanol, 99%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic drum
111-77-3
C5H12O3
120.15
SBASXUCJHJRPEV-UHFFFAOYSA-N
2-2-methoxyethoxy ethanol,diethylene glycol monomethyl ether,methyl carbitol,methyl digol,methoxydiglycol,ethanol, 2-2-methoxyethoxy,methyl dioxitol,diethylene glycol methyl ether,dowanol dm,poly-solv dm
8134
CHEBI:44836
2-(2-methoxyethoxy)ethanol
COCCOCCO
111-77-3
C5H12O3
120.15
SBASXUCJHJRPEV-UHFFFAOYSA-N
2-2-methoxyethoxy ethanol,diethylene glycol monomethyl ether,methyl carbitol,methyl digol,methoxydiglycol,ethanol, 2-2-methoxyethoxy,methyl dioxitol,diethylene glycol methyl ether,dowanol dm,poly-solv dm
8134
CHEBI:44836
2-(2-methoxyethoxy)ethanol
COCCOCCO
Viscosity 3.9 mPa.s (20°C)
Density 1.0100g/mL
Boiling Point 194.0°C
Color Undesignated
Packaging Glass Bottle
Melting Point -70.0°C
Quantity 1L
Flash Point 83°C
Infrared Spectrum Authentic
Refractive Index 1.4250 to 1.4270
Specific Gravity 1.01
Assay Percent Range 99%
Linear Formula CH3OCH2CH2OCH2CH2OH
Beilstein 01,IV,2392
Merck Index 15,3136
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,glycerol,ether,acetone,and dmf
Formula Weight 120.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-(2-Methoxyethoxy)ethanol
Show More Show Less

GHS P Statement
Suspected of damaging the unborn child.

GHS P Statement
Use personal protective equipment as required. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.