Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Methoxyethyl)pyridine 98.0+%, TCI America™

Click to view available options
Quantity:
25mL
114-91-0
C8H11NO
137.182
MFCD00023547
QRBVCAWHUSTDOT-UHFFFAOYSA-N
metyridine,2-2-methoxyethyl pyridine,methyridine,mintic,pyridine, 2-2-methoxyethyl,emthyridine,farmintic,methydrine,metiridin,prominthic
66991
CHEBI:81729
2-(2-methoxyethyl)pyridine
COCCC1=CC=CC=N1
114-91-0
C8H11NO
137.182
MFCD00023547
QRBVCAWHUSTDOT-UHFFFAOYSA-N
metyridine,2-2-methoxyethyl pyridine,methyridine,mintic,pyridine, 2-2-methoxyethyl,emthyridine,farmintic,methydrine,metiridin,prominthic
66991
CHEBI:81729
2-(2-methoxyethyl)pyridine
COCCC1=CC=CC=N1
Quantity 25mL
Boiling Point 96°C
Color Undesignated
Formula Weight 137.18
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-(2-Methoxyethyl)pyridine